MilliporeSigma
  • Induction of immunological tolerance to myelinogenic glial-restricted progenitor allografts.
Induction of immunological tolerance to myelinogenic glial-restricted progenitor allografts.
Brain : a journal of neurology (2019-09-19)
Shen Li, Byoung Chol Oh, Chengyan Chu, Antje Arnold, Anna Jablonska, Georg J Furtmüller, Hua-Min Qin, Johannes Boltze, Tim Magnus, Peter Ludewig, Mirosław Janowski, Gerald Brandacher, Piotr Walczak
ABSTRACT
The immunological barrier currently precludes the clinical utilization of allogeneic stem cells. Although glial-restricted progenitors have become attractive candidates to treat a wide variety of neurological diseases, their survival in immunocompetent recipients is limited. In this study, we adopted a short-term, systemically applicable co-stimulation blockade-based strategy using CTLA4-Ig and anti-CD154 antibodies to modulate T-cell activation in the context of allogeneic glial-restricted progenitor transplantation. We found that co-stimulation blockade successfully prevented rejection of allogeneic glial-restricted progenitors from immunocompetent mouse brains. The long-term engrafted glial-restricted progenitors myelinated dysmyelinated adult mouse brains within one month. Furthermore, we identified a set of plasma miRNAs whose levels specifically correlated to the dynamic changes of immunoreactivity and as such could serve as biomarkers for graft rejection or tolerance. We put forward a successful strategy to induce alloantigen-specific hyporesponsiveness towards stem cells in the CNS, which will foster effective therapeutic application of allogeneic stem cells.
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig1 Antibody, Chemicon®, from rabbit