Merck
  • Antihypertensive effects of exercise involve reshaping of gut microbiota and improvement of gut-brain axis in spontaneously hypertensive rat.
Antihypertensive effects of exercise involve reshaping of gut microbiota and improvement of gut-brain axis in spontaneously hypertensive rat.
Gut microbes (2021-01-01)
Wen-Jie Xia, Meng-Lu Xu, Xiao-Jing Yu, Meng-Meng Du, Xu-Hui Li, Tao Yang, Lu Li, Ying Li, Kai B Kang, Qing Su, Jia-Xi Xu, Xiao-Lian Shi, Xiao-Min Wang, Hong-Bao Li, Yu-Ming Kang
ABSTRACT
Exercise (Ex) has long been recognized to produce beneficial effects on hypertension (HTN). This coupled with evidence of gut dysbiosis and an impaired gut-brain axis led us to hypothesize that reshaping of gut microbiota and improvement in impaired gut-brain axis would, in part, be associated with beneficial influence of exercise. Male spontaneously hypertensive rats (SHR) and Wistar Kyoto (WKY) rats were randomized into sedentary, trained, and detrained groups. Trained rats underwent moderate-intensity exercise for 12 weeks, whereas, detrained groups underwent 8 weeks of moderate-intensity exercise followed by 4 weeks of detraining. Fecal microbiota, gut pathology, intestinal inflammation, and permeability, brain microglia and neuroinflammation were analyzed. We observed that exercise training resulted in a persistent decrease in systolic blood pressure in the SHR. This was associated with increase in microbial α diversity, altered β diversity, and enrichment of beneficial bacterial genera. Furthermore, decrease in the number of activated microglia, neuroinflammation in the hypothalamic paraventricular nucleus, improved gut pathology, inflammation, and permeability were also observed in the SHR following exercise. Interestingly, short-term detraining did not abolish these exercise-mediated improvements. Finally, fecal microbiota transplantation from exercised SHR into sedentary SHR resulted in attenuated SBP and an improved gut-brain axis. These observations support our concept that an impaired gut-brain axis is linked to HTN and exercise ameliorates this impairment to induce antihypertensive effects.
Sigma-Aldrich
Pyridinium dichromate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Methyl-L-tryptophan, 95%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Nystose, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Metronidazole, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Supelco
Vancomycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material