MilliporeSigma
  • TNF-alpha is the critical mediator of the cyclic AMP-induced apoptosis of CD8+4+ double-positive thymocytes.
TNF-alpha is the critical mediator of the cyclic AMP-induced apoptosis of CD8+4+ double-positive thymocytes.
Journal of immunology (Baltimore, Md. : 1950) (2000-02-05)
J A Guevara Patiño, V N Ivanov, E Lacy, K B Elkon, M W Marino, J Nikolic-Zugić
ABSTRACT
Apoptosis is one of the key regulatory mechanisms in tissue modeling and development. In the thymus, 95-98% of all thymocytes die by apoptosis because they failed to express a TCR with an optimal affinity for the selecting intrathymic peptide-MHC complexes. We studied the possible role of two prominent nerve growth factor (NGF-TNF) family member systems, Fas ligand (FasL)-Fas receptor (FasR) and TNF-alpha-TNFR, in apoptosis of murine CD8+4+ double-positive (DP) thymocytes induced via TCR-CD3- and cAMP-mediated signaling. TCR-CD3epsilon-mediated apoptosis of DP thymocytes was found not to be dependent on either of the two systems. The FasL-FasR system was also found to be dispensable for the cAMP-mediated apoptosis. By contrast, cAMP agonists (dibutyryl-cAMP and forskolin) induced apoptosis via TNF-alpha, as evidenced by 1) the ability of anti-TNF-alpha mAbs to abrogate cAMP analogue-induced DP apoptosis in a dose-dependent manner; and 2) increased resistance of DP thymocytes from TNF-alpha-/- and TNFR I-/-II-/- animals to cAMP agonist-mediated apoptosis. cAMP agonists induced DP thymocyte death by a combination of two mechanisms: first, they induced selective up-regulation of TNF-alpha production, and, second, they sensitized DP thymocytes to TNF-alpha. The latter effect may be due to the down-regulation of TNFR-associated factor 2 protein. These results identify TNF-alpha as the critical mediator of cAMP-induced apoptosis in thymocytes and provide a molecular explanation for how the cAMP stimulators, including the sex steroids, may modulate T cell production output, as observed under physiological and pharmacological conditions.
Sigma-Aldrich
Anti-TNFα Antibody, clone MPG-XT3, clone MPG-XT3, Upstate®, from rat