Merck
Off-rate screening for selection of high-affinity anti-drug antibodies.
Analytical biochemistry (2013-08-03)
Francisco Ylera, Stefan Harth, Dirk Waldherr, Christian Frisch, Achim Knappik
ABSTRACT
The rapidly increasing number of therapeutic antibodies in clinical development and on the market requires corresponding detection reagents for monitoring the concentration of these drugs in patient samples and as positive controls for measurement of anti-drug antibodies. Phage display of large recombinant antibody libraries has been shown to enable the rapid development of fully human anti-idiotypic antibodies binding specifically to antibody drugs, since the in vitro panning approach allows for incorporation of suitable blockers to drive selection toward the paratope of the drug. A typical bottleneck in antibody generation projects is ranking of the many candidates obtained after panning on the basis of antibody binding strength. Ideally, such method will work without prior labeling of antigens and with crude bacterial lysates. We developed an off-rate screening method of crude Escherichia coli lysates containing monovalent Fab fragments obtained after phage display of the HuCAL PLATINUM® antibody library. We used the antibody drugs trastuzumab and cetuximab as antigen examples. Using the Octet® RED384 label-free sensor instrument we show that antibody off rates can be reliably determined in crude bacterial lysates with high throughput. We also demonstrate that the method can be applied to screening for high-affinity antibodies typically obtained after affinity maturation.
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered