MilliporeSigma
  • Synergy between polyethylenimine and different families of antibiotics against a resistant clinical isolate of Pseudomonas aeruginosa.
Synergy between polyethylenimine and different families of antibiotics against a resistant clinical isolate of Pseudomonas aeruginosa.
Antimicrobial agents and chemotherapy (2008-02-21)
Hayssam Khalil, Tao Chen, Renée Riffon, Rutao Wang, Zhao Wang
ABSTRACT
The in vitro activities of 10 families of antimicrobial agents alone and in combination with a synthetic polycationic polymer, polyethylenimine (PEI), against a resistant clinical isolate of Pseudomonas aeruginosa were investigated by MIC assays, checkerboard testing, and killing curve studies. At a concentration of 250 nM, PEI (10 kDa) was not directly bactericidal or bacteriostatic; but when it was used in combination with novobiocin, ceftazidime, ampicillin, ticarcillin, carbenicillin, piperacillin, cefotaxime, chloramphenicol, rifampin, or norfloxacin, it significantly reduced the MICs of these antibiotics by 1.5- to 56-fold. However, the MICs of aminoglycosides, polymyxins, and vancomycins were increased by 1.2- to 5-fold when these drugs were combined with PEI; and the MICs of tetracycline, erythromycin, ciprofloxacin, and ofloxacin were not affected when these drugs were combined with PEI. In the killing curve studies, combinations of PEI with novobiocin, ceftazidime, chloramphenicol, or rifampin resulted in 5- to 8-log(10) CFU/ml reductions in bacterial counts when 25% of the MIC of each antibiotic was used. These results indicate that infections due to resistant Pseudomonas strains could be treated by the use of a synergistic combination of PEI and antimicrobial drugs.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)