- Whole-Body but Not Hepatic Knockdown of Chemerin by Antisense Oligonucleotide Decreases Blood Pressure in Rats.
Whole-Body but Not Hepatic Knockdown of Chemerin by Antisense Oligonucleotide Decreases Blood Pressure in Rats.
Chemerin is an inflammatory adipokine positively associated with hypertension and obesity. The majority of chemerin derives from the liver and adipose tissue, however, their individual contributions to blood pressure are unknown. We began studying chemerin in the normal rat using antisense oligonucleotides (ASO) with whole-body activity (Gen 2.5 chemerin ASO) or liver-restricted activity (GalNAc chemerin ASO). We hypothesized that in normotensive male Sprague-Dawley rats, circulating chemerin is predominately liver-derived and regulates blood pressure. A dosing study of the Gen 2.5 chemerin ASO (with a scrambled control ASO) supported 25 mg/kg as the appropriate dose. GalNAc chemerin ASO was also assessed and used at 10 mg/kg. Radiotelemetry monitored mean arterial pressure (MAP) for a 1-week baseline and weekly subcutaneous ASO injections for 4 weeks. Two days after the final injection, animals were euthanized for tissue reverse transcription-polymerase chain reaction and chemerin Western analysis. Gen 2.5 chemerin ASO treatments reduced chemerin mRNA and protein in liver, retroperitoneal fat (RP), and mesenteric perivascular adipose tissue (mPVAT), as well as reducing protein in plasma. GalNAc chemerin ASO treatments reduced chemerin mRNA and protein in liver and chemerin protein in plasma but had no effect on expression in RP fat or mPVAT. Gen 2.5 chemerin ASO treatment reduced MAP compared with control ASO but was unchanged in animals receiving the GalNAc chemerin ASO. Although circulating chemerin is liver-derived, it does not play a major role in blood pressure regulation. Local effects of chemerin from fat may explain this discrepancy and support chemerin's association with hypertension and obesity.