MilliporeSigma
  • Triptolide sensitizes pancreatic cancer cells to TRAIL-induced activation of the death receptor pathway.
Triptolide sensitizes pancreatic cancer cells to TRAIL-induced activation of the death receptor pathway.
Cancer letters (2014-03-26)
Zhiyu Chen, Veena Sangwan, Sulagna Banerjee, Rohit Chugh, Vikas Dudeja, Selwyn M Vickers, Ashok K Saluja
ABSTRACT
The tumor necrosis factor related apoptosis-inducing ligand (TRAIL) causes cancer cell death, but many cancers, including pancreatic cancer, are resistant to TRAIL therapy. A combination of TRAIL and the diterpene triepoxide, triptolide, is effective in inducing pancreatic cancer cell death. Triptolide increases levels of death receptor DR5 and decreases the pro-survival FLICE-like inhibitory protein (c-FLIP), which contribute to the activation of caspase-8. This combination further causes both lysosomal and mitochondrial membrane permeabilization, resulting in cell death. Our study provides a mechanism by which triptolide sensitizes TRAIL resistant cells, which may become a novel therapeutic strategy against pancreatic cancer.
Sigma-Aldrich
Anti-Cathepsin B (Ab-1) Mouse mAb (CA10), lyophilized, clone CA10, Calbiochem®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00