MilliporeSigma
Chemical Constituents of the Roots of Euphorbia micractina.
Journal of natural products (2009-08-26)
Wendong Xu, Chenggen Zhu, Wei Cheng, Xiaona Fan, Xiaoguang Chen, Sen Yang, Ying Guo, Fei Ye, Jiangong Shi
ABSTRACT
Nine minor new tirucallane (1-7) and euphane (8 and 9) triterpenoids including five hydroperoxides, together with 18 known compounds, have been isolated from an ethanolic extract of the roots of Euphorbia micractina. Their structures including absolute configurations were elucidated by spectroscopic and chemical analysis. In the in vitro assays, betulin (10) showed a selective cytotoxic activity against A2780 ovarian cells with an IC(50) value of 6.1 microM and inhibitory activity against protein tyrosine phosphatase 1B (PTP1B) with an IC(50) value of 15.3 microM. Jolkinol B (11) showed a potent activity against HIV-1 replication with an IC(50) value of 12.6 muM. However, compounds 1-9 and the other known compounds were inactive in the three assays used.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-α-Palmitin, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Scopoletin, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleanolic acid, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Betulin, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Cinnamic acid, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Sitosterol, ≥70%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform