MilliporeSigma
  • Roflupram, a novel phosphodiesterase 4 inhibitor, inhibits lipopolysaccharide-induced neuroinflammatory responses through activation of the AMPK/Sirt1 pathway.
Roflupram, a novel phosphodiesterase 4 inhibitor, inhibits lipopolysaccharide-induced neuroinflammatory responses through activation of the AMPK/Sirt1 pathway.
International immunopharmacology (2020-11-28)
Dan Li, Jiangping Xu, Yunyun Qin, Ningbo Cai, Yufang Cheng, Haitao Wang
ABSTRACT
Roflupram (ROF) is a novel phosphodiesterase 4 inhibitor. We previously found that ROF suppressed the production of pro-inflammatory factors in microglial cells; however, the underlying mechanisms are largely unknown. The present study aimed to elucidate the underlying molecular mechanisms of the anti-neuroinflammatory effects of ROF in lipopolysaccharide (LPS)-activated microglial cells and LPS-challenged mice. Treatment with ROF suppressed LPS-induced expression of interleukin (IL)-6 and tumor necrosis factor (TNF)-α in BV-2 microglia cell line. Immunofluorescence and Western blotting analysis showed that ROF significantly inhibited the activation of microglia, as evidenced by decreased expression of ionized calcium binding adaptor molecule-1 (Iba1). Similar results were obtained in primary cultured microglial cells. ROF induced the phosphorylation of AMP-activated protein kinase (AMPK) and the expression of Sirtuin 1 (Sirt1). Interestingly, the AMPK inhibitor, compound C, blocked the role of ROF in both the phosphorylation of AMPK and the expression of Sirt1 in BV-2 cells stimulated with LPS. More importantly, the Sirt1 inhibitor, EX527, abolished the inhibitory role of ROF on the production of pro-inflammatory factors, and reactivated BV-2 cells. In mice challenged with LPS, ROF improved cognition and decreased the levels of IL-6 and TNF-α in both the cortex and hippocampus. In contrast, EX527 weakened the effects of ROF on cognitive enhancement and reduction of pro-inflammatory factors in the cortex and hippocampus. Furthermore, EX527 blocked the inhibitory role of ROF in the activation of microglial cells in both the hippocampus and cortex. Taken together, our results indicated that ROF attenuated LPS-induced neuroinflammatory responses in microglia, and the AMPK/Sirt1 pathway is essential for the anti-inflammatory effects of ROF.
Sigma-Aldrich
Roflupram, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PDE4B
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)