MilliporeSigma
  • Synthesis, antiproliferative activity and inhibition of tubulin polymerization by anthracenone-based oxime derivatives.
Synthesis, antiproliferative activity and inhibition of tubulin polymerization by anthracenone-based oxime derivatives.
European journal of medicinal chemistry (2010-06-16)
Georg Surkau, Konrad J Böhm, Klaus Müller, Helge Prinz
ABSTRACT
A series of anthracenone-based oxime ethers and -esters were synthesized in order to evaluate their antiproliferative activity. Several investigated compounds displayed strong antiproliferative activity against K562 leukemia cells and proved to be strong inhibitors of tubulin polymerization. In this context, anthracenone-based oxime ethers and -esters are considered to contribute to the development of novel antiproliferative drugs, based on tubulin interaction.
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Podophyllotoxin
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)