MilliporeSigma
  • Furostanol saponins from the rhizomes of Dioscorea japonica and their effects on NGF induction.
Furostanol saponins from the rhizomes of Dioscorea japonica and their effects on NGF induction.
Bioorganic & medicinal chemistry letters (2011-03-01)
Ki Hyun Kim, Min Ah Kim, Eunjung Moon, Sun Yeou Kim, Sang Zin Choi, Mi Won Son, Kang Ro Lee
ABSTRACT
The rhizome of Dioscorea japonica is a food and medicinal source known as 'San Yak' in Korea. Two new furostanol saponins, coreajaponins A (1) and B (2), together with 10 known compounds (3-12) were isolated from the rhizomes of D. japonica. Their structures were determined by spectroscopic methods, including 1D and 2D NMR techniques, HRMS, and chemical methods. Nerve growth factor (NGF), a crucial factor for neuronal survival and differentiation, can potentially improve neurodegenerative diseases and diabetic polyneuropathy. We evaluated the effects of isolates (1-12) on NGF induction in a C6 rat glioma cell line. Coreajaponin B (2) upregulated NGF content without significant cell toxicity, as did 6, 8, 9, and 11.
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Sigma-Aldrich
β-Sitosterol, ≥70%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$183.00
2.5 L
Estimated to ship on March 21, 2025
$380.00