MilliporeSigma
  • DNA origami demonstrate the unique stimulatory power of single pMHCs as T cell antigens.
DNA origami demonstrate the unique stimulatory power of single pMHCs as T cell antigens.
Proceedings of the National Academy of Sciences of the United States of America (2021-01-21)
Joschka Hellmeier, Rene Platzer, Alexandra S Eklund, Thomas Schlichthaerle, Andreas Karner, Viktoria Motsch, Magdalena C Schneider, Elke Kurz, Victor Bamieh, Mario Brameshuber, Johannes Preiner, Ralf Jungmann, Hannes Stockinger, Gerhard J Schütz, Johannes B Huppa, Eva Sevcsik
ABSTRACT
T cells detect with their T cell antigen receptors (TCRs) the presence of rare agonist peptide/MHC complexes (pMHCs) on the surface of antigen-presenting cells (APCs). How extracellular ligand binding triggers intracellular signaling is poorly understood, yet spatial antigen arrangement on the APC surface has been suggested to be a critical factor. To examine this, we engineered a biomimetic interface based on laterally mobile functionalized DNA origami platforms, which allow for nanoscale control over ligand distances without interfering with the cell-intrinsic dynamics of receptor clustering. When targeting TCRs via stably binding monovalent antibody fragments, we found the minimum signaling unit promoting efficient T cell activation to consist of two antibody-ligated TCRs within a distance of 20 nm. In contrast, transiently engaging antigenic pMHCs stimulated T cells robustly as well-isolated entities. These results identify pairs of antibody-bound TCRs as minimal receptor entities for effective TCR triggering yet validate the exceptional stimulatory potency of single isolated pMHC molecules.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture