MilliporeSigma
  • Enhancement of the Mechanical and Drug-Releasing Properties of Poloxamer 407 Hydrogels with Casein.
Enhancement of the Mechanical and Drug-Releasing Properties of Poloxamer 407 Hydrogels with Casein.
Pharmaceutical research (2021-02-28)
Louise Lacalendola Tundisi, Rong Yang, Luiz Phellipe Pozzuto Borelli, Thais Alves, Manisha Mehta, Marco Vinícius Chaud, Priscila Gava Mazzola, Daniel S Kohane
ABSTRACT
Topical therapy of local disease (e.g. skin) is advantageous over oral therapy since there is less systemic drug distribution (so fewer side-effects), no first-pass effect, etc. However, patient compliance with topical therapy can be poor as it may require many applications a day and can last months. Here we propose a topical controlled release formulation with thermoresponsive gelation at body temperature and improved adhesiveness, making it easier to remain in contact with the body. The formulation contains two excipients, poloxamer 407 (P407) and casein. Casein can modify the properties of the hydrogel through molecular entanglement. In addition, tissue reaction and drug release profile were evaluated. Changes in casein concentration affected adhesive strength, viscosity, mechanical properties and drug release, presumably by hydrophobic interactions between casein and P407. Two different concentrations of P407 were tested with two different concentrations of casein. Formulations containing 5% and 10% casein released 80% of model drug in 48 h, while formulations without casein released the same fraction in around 24 h hours. Formulations with 10% casein had almost twice the adhesive strength of those without casein. Addition of casein modified the mechanical properties and drug release rate of the hydrogel. There was no inflammation or injury after brief exposure in vivo.
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Casein from bovine milk, powder