Merck
  • Regulation of m6A Methylation as a New Therapeutic Option against COVID-19.
Regulation of m6A Methylation as a New Therapeutic Option against COVID-19.
Pharmaceuticals (Basel, Switzerland) (2021-11-28)
Carla Zannella, Luca Rinaldi, Giovanni Boccia, Annalisa Chianese, Ferdinando Carlo Sasso, Francesco De Caro, Gianluigi Franci, Massimiliano Galdiero
ABSTRACT
The rapid spread of SARS-CoV-2 and the resulting pandemic has led to a spasmodic search for approaches able to limit the diffusion of the disease. The epigenetic machinery has aroused considerable interest in the last decades, and much evidence has demonstrated that this type of modification could regulate the early stages of viral infection. Recently it was reported that N6-methyladenosine (m6A) influences SARS-CoV-2 replication, although its role remains to be further investigated. The knockdown of enzymes involved in the m6A pathway could represent an optimal strategy to deepen the epigenetic mechanism. In the present study, we blocked the catalytic activity of the fat mass and obesity-associated protein (FTO) by using the selective inhibitor rhein. We observed a strong broad-spectrum reduction of infectivity caused by various coronaviruses, including SARS-CoV-2. This effect could be due to the modulation of m6A levels and could allow identification of this modification as a new therapeutic target to treat SARS-CoV-2 infection.
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rhein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%