MilliporeSigma
  • Determination of H+-ATPase Activity in Arabidopsis Guard Cell Protoplasts through H+-pumping Measurement and H+-ATPase Quantification.
Determination of H+-ATPase Activity in Arabidopsis Guard Cell Protoplasts through H+-pumping Measurement and H+-ATPase Quantification.
Bio-protocol (2017-12-20)
Shota Yamauchi, Ken-Ichiro Shimazaki
ABSTRACT
The opening of stomata in plants in response to blue light is driven by the plasma membrane H+-ATPase in guard cells. To evaluate the activation of the H+-ATPase in vivo, we can use H+-pumping by guard cells in response to blue light and fusicoccin. To do this, it is required to prepare a large amount of guard cell protoplasts and measure H+-pumping in the protoplasts. It is also necessary to determine the protein amount of H+-ATPase. In this protocol, we describe the procedures required for these preparations and measurements.
Sigma-Aldrich
Nα-Tosyl-Lys Chloromethyl Ketone, Hydrochloride, Inhibits trypsin-like serine proteinases.
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.