MilliporeSigma
  • Anti-stress constituents of Evolvulus alsinoides: an ayurvedic crude drug.
Anti-stress constituents of Evolvulus alsinoides: an ayurvedic crude drug.
Chemical & pharmaceutical bulletin (2007-05-03)
Prasoon Gupta, Akanksha, Kiran Babu Siripurapu, Ausaf Ahmad, Gautam Palit, Ashish Arora, Rakesh Maurya
ABSTRACT
Bioactivity-guided purification of n-BuOH soluble fraction from the ethanol extract of Evolvulus alsinoides resulted in the isolation of two new compounds, 2,3,4-trihydroxy-3-methylbutyl 3-[3-hydroxy-4-(2,3,4-trihydroxy-2-methylbutoxy)-phenyl]-2-propenoate (1) and 1,3-di-O-caffeoyl quinic acid methyl ester (2) along with six known compounds, caffeic acid (3), 6-methoxy-7-O-beta-glucopyranoside coumarin (4), 2-C-methyl erythritol (5), kaempferol-7-O-beta-glucopyranoside (6), kaempferol-3-O-beta-glucopyranoside (7) and quecetine-3-O-beta-glucopyranoside (8). The structure of new compounds 1 and 2 were elucidated by spectroscopic analysis, while known compounds were confirmed by direct comparison of their NMR data with those reported in literature. This is the first report of the presence of phenolic constituents in Evolvulus alsinoides. The isolated compounds 1-5 and 8 were screened for anti-stress activity in acute stress induced biochemical changes in adult male Sprague-Dawley rats. Stress exposure has resulted in significant increase of plasma glucose, adrenal gland weight, plasma creatine kinase (CK), and corticosterone levels. Compound 1 displayed most promising antistress effect by normalizing hyperglycemia, plasma corticosterone, CK and adrenal hypertrophy, while compounds 2 and 3 were also effective in normalizing most of these stress parameters, however compounds 4, 5 and 8 were ineffective in normalizing these parameters.
Sigma-Aldrich
Ginseng root from Panax quinquefolium (American ginseng)