MilliporeSigma
  • Parkin overexpression ameliorates hippocampal long-term potentiation and β-amyloid load in an Alzheimer's disease mouse model.
Parkin overexpression ameliorates hippocampal long-term potentiation and β-amyloid load in an Alzheimer's disease mouse model.
Human molecular genetics (2013-10-10)
Xiaoqi Hong, Jie Liu, Guoqi Zhu, YingHan Zhuang, Haiyun Suo, Pan Wang, Dongping Huang, Jing Xu, Yufang Huang, Mei Yu, MinJuan Bian, Zhejin Sheng, Jian Fei, Houyan Song, Thomas Behnisch, Fang Huang
ABSTRACT
Alzheimer's disease (AD) is a progressive neurodegenerative disorder characterized by a severe decline of memory performance. A widely studied AD mouse model is the APPswe/PSEN1ΔE9 (APP/PS1) strain, as mice exhibit amyloid plaques as well as impaired memory capacities. To test whether restoring synaptic plasticity and decreasing β-amyloid load by Parkin could represent a potential therapeutic target for AD, we crossed APP/PS1 transgenic mice with transgenic mice overexpressing the ubiquitin ligase Parkin and analyzed offspring properties. Overexpression of Parkin in APP/PS1 transgenic mice restored activity-dependent synaptic plasticity and rescued behavioral abnormalities. Moreover, overexpression of Parkin was associated with down-regulation of APP protein expression, decreased β-amyloid load and reduced inflammation. Our data suggest that Parkin could be a promising target for AD therapy.
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.