MilliporeSigma
  • A hotspot mutation targeting the R-RAS2 GTPase acts as a potent oncogenic driver in a wide spectrum of tumors.
A hotspot mutation targeting the R-RAS2 GTPase acts as a potent oncogenic driver in a wide spectrum of tumors.
Cell reports (2022-03-17)
Isabel Fernández-Pisonero, Laura Clavaín, Javier Robles-Valero, L Francisco Lorenzo-Martín, Rubén Caloto, Blanca Nieto, Carmen García-Macías, Clara L Oeste, Manuel Sánchez-Martín, Antonio Abad, Alejandro Hortal, Dolores Caballero, Marcos González, Mercedes Dosil, Balbino Alarcón, Xosé R Bustelo
ABSTRACT
A missense change in RRAS2 (Gln72 to Leu), analogous to the Gln61-to-Leu mutation of RAS oncoproteins, has been identified as a long-tail hotspot mutation in cancer and Noonan syndrome. However, the relevance of this mutation for in vivo tumorigenesis remains understudied. Here we show, using an inducible knockin mouse model, that R-Ras2Q72L triggers rapid development of a wide spectrum of tumors when somatically expressed in adult tissues. These tumors show limited overlap with those originated by classical Ras oncogenes. R-Ras2Q72L-driven tumors can be classified into different subtypes according to therapeutic susceptibility. Importantly, the most relevant R-Ras2Q72L-driven tumors are dependent on mTORC1 but independent of phosphatidylinositol 3-kinase-, MEK-, and Ral guanosine diphosphate (GDP) dissociation stimulator. This pharmacological vulnerability is due to the extensive rewiring by R-Ras2Q72L of pathways that orthogonally stimulate mTORC1 signaling. These findings demonstrate that RRAS2Q72L is a bona fide oncogenic driver and unveil therapeutic strategies for patients with cancer and Noonan syndrome bearing RRAS2 mutations.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
$47.90
25 g
In Stock
Details...
$125.00
100 g
In Stock
Details...
$319.00
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
$47.90
25 g
In Stock
Details...
$125.00
100 g
In Stock
Details...
$319.00
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
$47.90
25 g
In Stock
Details...
$125.00
100 g
In Stock
Details...
$319.00
Roche
Fibronectin, from human plasma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
$47.90
25 g
In Stock
Details...
$125.00
100 g
In Stock
Details...
$319.00