MilliporeSigma
  • Structural Basis of the Activation of Heterotrimeric Gs-Protein by Isoproterenol-Bound β1-Adrenergic Receptor.
Structural Basis of the Activation of Heterotrimeric Gs-Protein by Isoproterenol-Bound β1-Adrenergic Receptor.
Molecular cell (2020-08-21)
Minfei Su, Lan Zhu, Yixiao Zhang, Navid Paknejad, Raja Dey, Jianyun Huang, Ming-Yue Lee, Dewight Williams, Kelsey D Jordan, Edward T Eng, Oliver P Ernst, Joel R Meyerson, Richard K Hite, Thomas Walz, Wei Liu, Xin-Yun Huang
ABSTRACT
Cardiac disease remains the leading cause of morbidity and mortality worldwide. The β1-adrenergic receptor (β1-AR) is a major regulator of cardiac functions and is downregulated in the majority of heart failure cases. A key physiological process is the activation of heterotrimeric G-protein Gs by β1-ARs, leading to increased heart rate and contractility. Here, we use cryo-electron microscopy and functional studies to investigate the molecular mechanism by which β1-AR activates Gs. We find that the tilting of α5-helix breaks a hydrogen bond between the sidechain of His373 in the C-terminal α5-helix and the backbone carbonyl of Arg38 in the N-terminal αN-helix of Gαs. Together with the disruption of another interacting network involving Gln59 in the α1-helix, Ala352 in the β6-α5 loop, and Thr355 in the α5-helix, these conformational changes might lead to the deformation of the GDP-binding pocket. Our data provide molecular insights into the activation of G-proteins by G-protein-coupled receptors.
Sigma-Aldrich
4-(3-Butoxy-4-methoxybenzyl)imidazolidin-2-one, solid
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Roche
Adenosine Deaminase (ADA), from calf intestine
Sigma-Aldrich
(−)-Isoproterenol hydrochloride