MilliporeSigma
  • Ral GTPases are critical regulators of spinal cord myelination and homeostasis.
Ral GTPases are critical regulators of spinal cord myelination and homeostasis.
Cell reports (2022-09-29)
Jonathan DeGeer, Anna Lena Datwyler, Chiara Rickenbach, Andrea Ommer, Daniel Gerber, Cristina Fimiani, Joanne Gerber, Jorge A Pereira, Ueli Suter
ABSTRACT
Efficient myelination supports nerve conduction and axonal health throughout life. In the central nervous system, oligodendrocytes (OLs) carry out this demanding anabolic duty in part through biosynthetic pathways controlled by mTOR. We identify Ral GTPases as critical regulators of mouse spinal cord myelination and myelin maintenance. Ablation of Ral GTPases (RalA, RalB) in OL-lineage cells impairs timely onset and radial growth of developmental myelination, accompanied by increased endosomal/lysosomal abundance. Further examinations, including transcriptomic analyses of Ral-deficient OLs, were consistent with mTORC1-related deficits. However, deletion of the mTOR signaling-repressor Pten in Ral-deficient OL-lineage cells is unable to rescue mTORC1 activation or developmental myelination deficiencies. Induced deletion of Ral GTPases in OLs of adult mice results in late-onset myelination defects and tissue degeneration. Together, our data indicate critical roles for Ral GTPases to promote developmental spinal cord myelination, to ensure accurate mTORC1 signaling, and to protect the healthy state of myelin-axon units over time.
Sigma-Aldrich
Papain from papaya latex, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein