MilliporeSigma
  • Efficient transdifferentiation of human dermal fibroblasts into skeletal muscle.
Efficient transdifferentiation of human dermal fibroblasts into skeletal muscle.
Journal of tissue engineering and regenerative medicine (2017-01-20)
Selwa Mokhtar Boularaoui, Khaled M A Abdel-Raouf, Noaf Salah Ali Alwahab, Megan E Kondash, George A Truskey, Jeremy Choon Meng Teo, Nicolas Christoforou
ABSTRACT
Skeletal muscle holds significant regenerative potential but is incapable of restoring tissue loss caused by severe injury, congenital defects or tumour ablation. Consequently, skeletal muscle models are being developed to study human pathophysiology and regeneration. Their physiological accuracy, however, is hampered by the lack of an easily accessible human cell source that is readily expandable and capable of efficient differentiation. MYOD1, a master gene regulator, induces transdifferentiation of a variety of cell types into skeletal muscle, although inefficiently in human cells. Here we used MYOD1 to establish its capacity to induce skeletal muscle transdifferentiation of human dermal fibroblasts under baseline conditions. We found significant transdifferentiation improvement via transforming growth factor-β/activin signalling inhibition, canonical WNT signalling activation, receptor tyrosine kinase binding and collagen type I utilization. Mechanistically, manipulation of individual signalling pathways modulated the transdifferentiation process via myoblast proliferation, lowering the transdifferentiation threshold and inducing cell fusion. Overall, we used transdifferentiation to achieve the robust derivation of human skeletal myotubes and have described the signalling pathways and mechanisms regulating this process. Copyright © 2017 John Wiley & Sons, Ltd.
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Vimentin−Cy3 antibody produced in mouse, clone V9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fetuin from fetal bovine serum, lyophilized powder, suitable for cell culture