MilliporeSigma
  • Exploring the cytoskeleton during intracytoplasmic sperm injection in humans.
Exploring the cytoskeleton during intracytoplasmic sperm injection in humans.
Methods in molecular biology (Clifton, N.J.) (2008-12-17)
Vanesa Y Rawe, Héctor Chemes
ABSTRACT
Understanding the cellular events during fertilization in mammals is a major challenge that can contribute to the improvement of future infertility treatments in humans and reproductive performance in farm animals. Of special interest is the role of the oocyte and sperm cytoskeleton during the initial interaction between gametes. The aim of this chapter is to describe methods for studying cytoskeletal features during in vitro fertilization after intracytoplasmic sperm injection (ICSI) in humans. The following protocols will provide a detailed description of how to perform immunodetection and imaging of human eggs, zygotes, and sperm by fluorescence (confocal and epifluorescence) and electron microscopy.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sign Into View Organizational & Contract Pricing