MilliporeSigma
  • Spermine enhances antiviral and anticancer responses by stabilizing DNA binding with the DNA sensor cGAS.
Spermine enhances antiviral and anticancer responses by stabilizing DNA binding with the DNA sensor cGAS.
Immunity (2023-02-02)
Lina Wang, Siru Li, Kai Wang, Na Wang, Qiaoling Liu, Zhen Sun, Li Wang, Lulu Wang, Quentin Liu, Chengli Song, Qingkai Yang
ABSTRACT
Self-nonself discrimination is vital for the immune system to mount responses against pathogens while maintaining tolerance toward the host and innocuous commensals during homeostasis. Here, we investigated how indiscriminate DNA sensors, such as cyclic GMP-AMP synthase (cGAS), make this self-nonself distinction. Screening of a small-molecule library revealed that spermine, a well-known DNA condenser associated with viral DNA, markedly elevates cGAS activation. Mechanistically, spermine condenses DNA to enhance and stabilize cGAS-DNA binding, optimizing cGAS and downstream antiviral signaling. Spermine promotes condensation of viral, but not host nucleosome, DNA. Deletion of viral DNA-associated spermine, by propagating virus in spermine-deficient cells, reduced cGAS activation. Spermine depletion subsequently attenuated cGAS-mediated antiviral and anticancer immunity. Collectively, our results reveal a pathogenic DNA-associated molecular pattern that facilitates nonself recognition, linking metabolism and pathogen recognition.
Sigma-Aldrich
Dynamin Inhibitor I, Dynasore, The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.
SAFC
Spermidine trihydrochloride, ≥99.0% (AT)