MilliporeSigma
  • Measuring DNA modifications with the comet assay: a compendium of protocols.
Measuring DNA modifications with the comet assay: a compendium of protocols.
Nature protocols (2023-01-29)
Andrew Collins, Peter Møller, Goran Gajski, Soňa Vodenková, Abdulhadi Abdulwahed, Diana Anderson, Ezgi Eyluel Bankoglu, Stefano Bonassi, Elisa Boutet-Robinet, Gunnar Brunborg, Christy Chao, Marcus S Cooke, Carla Costa, Solange Costa, Alok Dhawan, Joaquin de Lapuente, Cristian Del Bo', Julien Dubus, Maria Dusinska, Susan J Duthie, Naouale El Yamani, Bevin Engelward, Isabel Gaivão, Lisa Giovannelli, Roger Godschalk, Sofia Guilherme, Kristine B Gutzkow, Khaled Habas, Alba Hernández, Oscar Herrero, Marina Isidori, Awadhesh N Jha, Siegfried Knasmüller, Ingeborg M Kooter, Gudrun Koppen, Marcin Kruszewski, Carina Ladeira, Blanca Laffon, Marcelo Larramendy, Ludovic Le Hégarat, Angélique Lewies, Anna Lewinska, Guillermo E Liwszyc, Adela López de Cerain, Mugimane Manjanatha, Ricard Marcos, Mirta Milić, Vanessa Moraes de Andrade, Massimo Moretti, Damian Muruzabal, Matjaž Novak, Rui Oliveira, Ann-Karin Olsen, Norah Owiti, Mário Pacheco, Alok K Pandey, Stefan Pfuhler, Bertrand Pourrut, Kerstin Reisinger, Emilio Rojas, Elise Rundén-Pran, Julen Sanz-Serrano, Sergey Shaposhnikov, Ville Sipinen, Karen Smeets, Helga Stopper, João Paulo Teixeira, Vanessa Valdiglesias, Mahara Valverde, Frederique van Acker, Frederik-Jan van Schooten, Marie Vasquez, Johannes F Wentzel, Maciej Wnuk, Annelies Wouters, Bojana Žegura, Tomas Zikmund, Sabine A S Langie, Amaya Azqueta
ABSTRACT
The comet assay is a versatile method to detect nuclear DNA damage in individual eukaryotic cells, from yeast to human. The types of damage detected encompass DNA strand breaks and alkali-labile sites (e.g., apurinic/apyrimidinic sites), alkylated and oxidized nucleobases, DNA-DNA crosslinks, UV-induced cyclobutane pyrimidine dimers and some chemically induced DNA adducts. Depending on the specimen type, there are important modifications to the comet assay protocol to avoid the formation of additional DNA damage during the processing of samples and to ensure sufficient sensitivity to detect differences in damage levels between sample groups. Various applications of the comet assay have been validated by research groups in academia, industry and regulatory agencies, and its strengths are highlighted by the adoption of the comet assay as an in vivo test for genotoxicity in animal organs by the Organisation for Economic Co-operation and Development. The present document includes a series of consensus protocols that describe the application of the comet assay to a wide variety of cell types, species and types of DNA damage, thereby demonstrating its versatility.
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Millipore
Proteinase K, Lyophilized, Highly active serine protease that exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA.
Sigma-Aldrich
Collagenase Type IV, Cls IV
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma