MilliporeSigma
  • NF1 loss of function as an alternative initiating event in pancreatic ductal adenocarcinoma.
NF1 loss of function as an alternative initiating event in pancreatic ductal adenocarcinoma.
Cell reports (2022-11-10)
Gopalakrishnan Ramakrishnan, Parash Parajuli, Pura Singh, Creighton Friend, Eric Hurwitz, Celine Prunier, Mohammed S Razzaque, Keli Xu, Azeddine Atfi
ABSTRACT
A long-standing question in the pancreatic ductal adenocarcinoma (PDAC) field has been whether alternative genetic alterations could substitute for oncogenic KRAS mutations in initiating malignancy. Here, we report that Neurofibromin1 (NF1) inactivation can bypass the requirement of mutant KRAS for PDAC pathogenesis. An in-depth analysis of PDAC databases reveals various genetic alterations in the NF1 locus, including nonsense mutations, which occur predominantly in tumors with wild-type KRAS. Genetic experiments demonstrate that NF1 ablation culminates in acinar-to-ductal metaplasia, an early step in PDAC. Furthermore, NF1 haploinsufficiency results in a dramatic acceleration of KrasG12D-driven PDAC. Finally, we show an association between NF1 and p53 that is orchestrated by PML, and mosaic analysis with double markers demonstrates that concomitant inactivation of NF1 and Trp53 is sufficient to trigger full-blown PDAC. Together, these findings open up an exploratory framework for apprehending the mechanistic paradigms of PDAC with normal KRAS, for which no effective therapy is available.
Sigma-Aldrich
Caerulein, ≥95% (HPLC)
Sigma-Aldrich
Fibroblast Growth Factor-Basic, FGF-Basic, from human, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture