MilliporeSigma
  • Approach to the synthesis of dolabelides A and B: fragment synthesis by tandem silylformylation-crotylsilylation.
Approach to the synthesis of dolabelides A and B: fragment synthesis by tandem silylformylation-crotylsilylation.
Organic letters (2003-09-12)
Darby R Schmidt, Peter K Park, James L Leighton
ABSTRACT
[structure: see text] A synthesis of the C(15)-C(30) fragment of Dolabelides A and B has been achieved. The recently developed asymmetric silane alcoholysis and tandem silylformylation-crotylsilylation reactions were used as the key steps to establish the C(23)-C(27) 1,5-syn-diol. In addition, the flexibility of this methodology has been demonstrated with an efficient synthesis of the C(24)-C(25) trisubstituted olefin.
Sigma-Aldrich
Crotyl alcohol, mixture of cis and trans, 96%