MilliporeSigma
  • 1H NMR studies on the hydrogen-bonding network in mono-altro-beta-cyclodextrin and its complex with adamantane-1-carboxylic acid.
1H NMR studies on the hydrogen-bonding network in mono-altro-beta-cyclodextrin and its complex with adamantane-1-carboxylic acid.
Carbohydrate research (2005-05-12)
Birgit Hakkarainen, Kahee Fujita, Stefan Immel, Lennart Kenne, Corine Sandström
ABSTRACT
The hydrogen-bond network in mono-altro-beta-cyclodextrin and in its inclusion complex with adamantane-1-carboxylic acid were investigated by (1)H NMR spectroscopy using the chemical shifts, temperature coefficients and vicinal coupling constants of the exchangeable hydroxy protons. The chemical shifts of the 3-OH signals indicated that the hydrogen-bond network between the 2-OH and 3-OH groups was disturbed not only on each side of the altrose residue, but also along the whole dextrin chain. Upon addition of adamantane-1-carboxylic acid, altrose underwent a conformational change from the (1)C(4) to the (O)S(2) form, allowing a more continuous belt of hydrogen bonding, as evidenced by the downfield shift experienced by the 3-OH proton signals of the glucose residues.
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%