MilliporeSigma
  • Tween 40-based precipitate production observed on modified chromogenic agar and development of biological identification kit for Malassezia species.
Tween 40-based precipitate production observed on modified chromogenic agar and development of biological identification kit for Malassezia species.
Medical mycology (2006-05-17)
Takamasa Kaneko, Koichi Makimura, Takashi Sugita, Hideyo Yamaguchi
ABSTRACT
We developed a simple identification kit for nine species of Malassezia (M. furfur, M. slooffiae, M. sympodialis, M. restricta, M. obtusa, M. globosa, M. pachydermatis, M. dermatis, and M. japonica) based on their biological features. This method utilizes Tween 40-based precipitate production on modified chromogenic agar (CHROMagar) Malassezia medium, growth on specific agars (Sabouraud's dextrose agar, Cremophor EL agar, Tween 60-esculin agar), and catalase reactions. This identification kit was verified with 11 type and reference strains of nine Malassezia species. An additional 26 clinical isolates were also successfully identified using the kit and the results were confirmed by molecular biological analysis.
Sigma-Aldrich
ECO TWEEN® 40, viscous liquid
Sigma-Aldrich
TWEEN® 40, viscous liquid