MilliporeSigma
  • Preferential intramolecular ring closure of aminoalcohols with diethyl carbonate to oxazolidinones.
Preferential intramolecular ring closure of aminoalcohols with diethyl carbonate to oxazolidinones.
Chemical & pharmaceutical bulletin (2007-05-03)
Fumiko Fujisaki, Marumi Oishi, Kunihiro Sumoto
ABSTRACT
The closure by cyclization with diethyl carbonate (EtO)(2)CO from aminoalcohols 1 as starting material can lead to the oxazolidinones 2a, b and 2c, respectively. In the reaction of trans-isomer (6) and (EtO)(2)CO, isolated products were also only 5-membered oxazolidinone derivative (7), containing its dehydrated derivative 8. The preferential formation of the 5-membered oxazolidinone ring system apparently indicated that this process (5-Exo-Trig ring closure) is more favorable than that of 6- or 7-membered ring derivative (3 or 9) by 6- or 7-Exo-Trig ring closure.
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Diethyl carbonate, 99%