MilliporeSigma
  • Deep-ultraviolet solar-blind photoconductivity of individual gallium oxide nanobelts.
Deep-ultraviolet solar-blind photoconductivity of individual gallium oxide nanobelts.
Nanoscale (2011-01-05)
Liang Li, Erwin Auer, Meiyong Liao, Xiaosheng Fang, Tianyou Zhai, Ujjal K Gautam, Alois Lugstein, Yasuo Koide, Yoshio Bando, Dmitri Golberg
ABSTRACT
We designed solar-blind deep-ultraviolet semiconductor photodetectors using individual Ga2O3 nanobelts. The photoconductive behavior was systematically studied. The photodetectors demonstrate high selectivity towards 250 nm light, fast response times of less than 0.3 s, and a large photocurrent to dark current ratio of up to 4 orders of magnitude. The photoresponse parameters such as photocurrent, response time, and quantum efficiency depend strongly on the intensity of light, the detector environment, and the nanobelt size. The photoresponse mechanism was discussed, which was mainly attributed to the band bending, surface traps, and distribution of traps in the bandgap. Present Ga2O3 nanobelts can be exploited for future applications in photo sensing, light-emitting diodes, and optical switches.
Sigma-Aldrich
Gallium(III) oxide, ≥99.99% trace metals basis