MilliporeSigma
  • Preparation of chitin butyrate by using phosphoryl mixed anhydride system.
Preparation of chitin butyrate by using phosphoryl mixed anhydride system.
Carbohydrate research (2011-03-01)
Lok Ranjan Bhatt, Bo Mi Kim, Kim Hyun, Kyung Hee Kang, Chichong Lu, Kyu Yun Chai
ABSTRACT
Acylation of chitin with butyric acid was performed in the presence of trifluoroacetic anhydride/phosphoric acid mediated system. The products were characterized by (1)H NMR and FT-IR spectroscopy and their solubility was tested in different organic solvents. Inclusion of butyric acid moieties into the parent molecule was confirmed from the (1)H NMR and FT-IR spectra. FT-IR analysis revealed that the degree of acid substitution (DS) of the products was in a range of 1.9-2.38, which increased with increasing the amounts of butyric acid added to the reaction system. Degree of N-deacetylation (DD) of the products, as determined by (1)H NMR was between 54.2% and 65.6%. The products with DS >2.0 were soluble in dimethyl sulfoxide, N,N-dimethylformamide, tetrahydrofuran, methanol, acetone, chloroform, and acetic acid.
Product Description
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%