MilliporeSigma
  • Concentration and fractionation by isoelectric trapping in a micropreparative-scale multicompartmental electrolyzer having orthogonal pH gradients. Part 2.
Concentration and fractionation by isoelectric trapping in a micropreparative-scale multicompartmental electrolyzer having orthogonal pH gradients. Part 2.
Electrophoresis (2011-06-08)
Peniel J Lim, Gyula Vigh
ABSTRACT
A micropreparative-scale multicompartmental electrolyzer called ConFrac has been developed and tested for isoelectric trapping separations. ConFrac can be operated in pass-by-pass mode or recirculating mode, using either asymmetrical feeding (feed enters only the anodic or the cathodic flow-through compartment) or symmetrical feeding (feed enters both the anodic and the cathodic flow-through compartment). Symmetrical feeding results in higher processing rates and is the preferred operation mode. Residence time in the flow-through compartments is set as a compromise between processing rate and temperature rise in the effluent. Ampholytic components have been isolated from 5 to 50 mL volumes of micromolar feed solutions and hundredfold concentrated into 100-μL collection compartments. Samples containing ampholytic analytes in highly conducting salt solutions were readily desalted and fractionated in ConFrac in one operation. pH transients formerly observed in other isoelectric trapping devices were observed in ConFrac as well. The pH transients were caused by the unequal ion transmission rates of the anodic- and cathodic-side buffering membranes.
Sigma-Aldrich
Benzyltrimethylammonium bromide, 97%
Sigma-Aldrich
Silver p-toluenesulfonate, ≥99%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%
Sigma-Aldrich
Benzyltrimethylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium p-toluenesulfonate, 95%
Sigma-Aldrich
Benzyltrimethylammonium hydroxide solution, 40 wt. % in methanol
Sigma-Aldrich
Benzyltrimethylammonium chloride, 97%
Sigma-Aldrich
Benzyltrimethylammonium chloride solution, technical, ~60% in H2O
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, SAJ first grade, ≥99.0%