MilliporeSigma
Validation study on avermectine residues in foodstuffs.
Analytica chimica acta (2011-07-12)
L Giannetti, A Giorgi, F Necci, G Ferretti, F Buiarelli, B Neri
ABSTRACT
Avermectines are antiparasitic agents widely used as veterinary drugs for food producing animals. The European Community, due to their side effects, limited the use of these molecules establishing maximum residue limits (MRLs) in some foods. A validated qualitative and quantitative high performance liquid chromatography method with fluorescence detection (HPLC-FL) is presented for the simultaneous determination of ivermectin (IVM), abemectin (ABA), moxidectin (MOX), eprinomectin (EPR), doramectin (DOR) and emamectin (EMA) in foodstuffs (muscle, eggs and milk). Samples were extracted with acetonitrile, purified with liquid-liquid extraction (LLE), and analysed by HLPC-FL previous derivatization with trifluoroacetic anhydride (TFAA) in presence of 1-methyl-imidazole (MI) and acetic acid. To date, the presented method is the first validated for the matrix eggs, and in accordance with the requirements set by Commission Decision 2002/657/EC. Recoveries of the methods, calculated spiking the samples in the range 5.0-100.0 μg kg(-1), were 64-83% for muscle, 65-89% for milk and 63-84% for eggs. The precision (CV) ranged between 9.2 and 17.1% for muscle, 9.9 and 16.6% for milk and from 9.4 to 17.4% for eggs. Linearity for the six analytes was calculated from 5.0 to 200.0 μg kg(-1). The main advantages of the presented method are its rapidity, the specificity, the good precision and recovery that make it very suitable to the detection and determination of avermectines.
Product Description
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%