MilliporeSigma
  • Controlled synthesis of transition metal/conducting polymer nanocomposites.
Controlled synthesis of transition metal/conducting polymer nanocomposites.
Nanotechnology (2012-07-31)
Zhen Liu, Yang Liu, Lin Zhang, Selcuk Poyraz, Ning Lu, Moon Kim, James Smith, Xiaolong Wang, Yajiao Yu, Xinyu Zhang
ABSTRACT
A novel displacement reaction has been observed to occur between conducting polymers (CP) and metal salts which can be used to fabricate nanostructured CP-metal composites in a one-pot manner. Vanadium pentoxide (V(2)O(5)) nanofiber is used during the synthesis as the reactive seeds to induce the nanofibril CP-metal network formation. The CP-metal nanocomposites exhibit excellent sensory properties for hydrogen peroxide (H(2)O(2)) detection, where both high sensitivity and a low detection limit can be obtained. The sensory performance of the CP-metal composite can be further enhanced by a facile microwave treatment. It is believed that the CP-metal nanofibril network can be converted to a carbon-metal network by a microwave-induced carbonization process and result in the sensory enhancement.
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Vanadium(V) oxide, ≥99.6% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Vanadium(V) oxide, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Vanadium(V) oxide, ≥98%
Sign Into View Organizational & Contract Pricing