MilliporeSigma
  • Cosmetics as a potential source of environmental contamination in the UK.
Cosmetics as a potential source of environmental contamination in the UK.
Environmental technology (2012-09-20)
Danelle Dhanirama, Jan Gronow, Nikolaos Voulvoulis
ABSTRACT
Chemicals of emerging concern (CECs) are frequently used in cosmetic formulations and can potentially reach the environment at concentrations that may cause harm. A methodology was developed to assess over 120 chemicals assembled from product ingredient listings to identify and validate potential CECs in cosmetics, based on Annex XIII of REACH legislation. Ten potential CECs were identified: polydimethylsiloxane, butylated hydroxylanisole (BHA), butylated hydroxytoluene, triclosan, nano titanium dioxide, nano zinc oxide, butylparaben, diethyl phthalate, octinoxate methoxycinnamate and benzophenone. These chemicals were quantified based on their consumption and concentrations in cosmetics and percentage market penetration. The initial predicted environmental concentrations (PEC initial) were estimated to determine their exposure to the environment. With the exception of BHA, the PEC initial highlighted levels of exposure to the environment that triggered the need for further investigation of the chemicals. These chemicals were linked to cosmetics to highlight products with the potential to cause environmental harm. Skin care products had the highest quantities of CECs, with titanium dioxide and zinc oxide nanomaterials being dominant potential contaminants. Further research is required to assess the exposure pathways and fate of these chemicals to determine environmental risks associated with their use and disposal.
Product Description
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Diethyl phthalate, SAJ special grade, ≥98.0%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Benzophenone, European Pharmacopoeia (EP) Reference Standard