Merck
  • Hydrogen sulfide as an allosteric modulator of ATP-sensitive potassium channels in colonic inflammation.
Hydrogen sulfide as an allosteric modulator of ATP-sensitive potassium channels in colonic inflammation.
Molecular pharmacology (2012-11-02)
Aravind R Gade, Minho Kang, Hamid I Akbarali
ABSTRACT
The ATP-sensitive potassium channel (K(ATP)) in mouse colonic smooth muscle cell is a complex containing a pore-forming subunit (Kir6.1) and a sulfonylurea receptor subunit (SUR2B). These channels contribute to the cellular excitability of smooth muscle cells and hence regulate the motility patterns in the colon. Whole-cell voltage-clamp techniques were used to study the alterations in K(ATP) channels in smooth muscle cells in experimental colitis. Colonic inflammation was induced in BALB/C mice after intracolonic administration of trinitrobenzene sulfonic acid. K(ATP) currents were measured at a holding potential of -60 mV in high K(+) external solution. The concentration response to levcromakalim (LEVC), a K(ATP) channel opener, was significantly shifted to the left in the inflamed smooth-muscle cells. Both the potency and maximal currents induced by LEVC were enhanced in inflammation. The EC(50) values in control were 6259 nM (n = 10) and 422 nM (n = 8) in inflamed colon, and the maximal currents were 9.9 ± 0.71 pA/pF (60 μM) in control and 39.7 ± 8.8 pA/pF (3 μM) after inflammation. As was seen with LEVC, the potency and efficacy of sodium hydrogen sulfide (NaHS) (10-1000 μM) on K(ATP) currents were significantly greater in inflamed colon compared with controls. In control cells, pretreatment with 100 µM NaHS shifted the EC(50) for LEV-induced currents from 2838 (n = 6) to 154 (n = 8) nM. Sulfhydration of sulfonylurea receptor 2B (SUR2B) was induced by NaHS and colonic inflammation. These data suggest that sulfhydration of SUR2B induces allosteric modulation of K(ATP) currents in colonic inflammation.
Sigma-Aldrich
Cromakalim