MilliporeSigma
  • Pyrrole versus quinoline formation in the palladium catalyzed reaction of 2-alkynyl-3-bromothiophenes and 2-alkynyl-3-bromofurans with anilines. A combined experimental and computational study.
Pyrrole versus quinoline formation in the palladium catalyzed reaction of 2-alkynyl-3-bromothiophenes and 2-alkynyl-3-bromofurans with anilines. A combined experimental and computational study.
Organic & biomolecular chemistry (2012-11-07)
Ghazwan Ali Salman, Riffat Un Nisa, Viktor O Iaroshenko, Jamshed Iqbal, Khurshid Ayub, Peter Langer
ABSTRACT
Benzofuroquinolines were prepared by a new type of Pd catalyzed annulation reaction. In the first step, 2-alkynyl-3-bromobenzofurans were prepared by Sonogashira reactions of 2,3-dibromobenzofuran. Their Pd catalyzed reaction with electron-rich anilines afforded benzofuroquinolines by a domino C-N coupling/annulation process. This reaction proceeds as a C,N-cyclization via the nitrogen atom and the ortho-carbon of the aniline. Similarly, furoquinolines were prepared from 2,3-dibromofuran. In contrast, benzofuropyrroles and furopyrroles were formed by N,N-cyclization when electron-poor anilines were used. Earlier, we reported results related to the thiophene and benzothiophene series. Quinolines were formed from 2,3-dibromobenzothiophene when electron rich anilines were used. In contrast, pyrroles were obtained in the case of electron-poor anilines. On the other hand, pyrroles were generally obtained, not depending on the type of aniline, when 2,3-dibromothiophene was employed as the starting material. In the present article, a detailed DFT study related to the mechanism (quinoline versus pyrrole formation) is reported which provides a rationalization of the selectivities observed for the furan, benzofuran, thiophene and benzothiophene series and for the different selectivities observed for electron-rich and -poor anilines.
Product Description
Supelco
Aniline, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 24, 2025
$27.80
5 mL
Estimated to ship on March 24, 2025
$84.40
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
In Stock
Details...
$93.10
2.5 L
In Stock
Details...
$233.00
18 L
Estimated to ship on March 24, 2025
$667.00
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$316.00
$63.20
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
Estimated to ship on March 24, 2025
$39.20
100 mL
Estimated to ship on March 24, 2025
$49.50
500 mL
Estimated to ship on March 24, 2025
$118.00
4 x 100 mL
Only 0 left in stock
$120.00
Sigma-Aldrich
Bromine water, CP
Pricing and availability is not currently available.
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00
Sigma-Aldrich
Bromine, reagent grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$97.40
5 g
In Stock
Details...
$340.00