MilliporeSigma
  • Isolation, identification and antimicrobial activities of two secondary metabolites of Talaromyces verruculosus.
Isolation, identification and antimicrobial activities of two secondary metabolites of Talaromyces verruculosus.
Molecules (Basel, Switzerland) (2012-11-30)
Fang Miao, Rui Yang, Dong-Dong Chen, Ying Wang, Bao-Fu Qin, Xin-Juan Yang, Le Zhou
ABSTRACT
From the ethyl acetate extract of the culture broth of Talaromyces verruculosus, a rhizosphere fungus of Stellera chamaejasme L., (-)-8-hydroxy-3-(4-hydroxypentyl)-3,4-dihydroisocoumarin (1) and (E)-3-(2,5-dioxo-3-(propan-2-ylidene)pyrrolidin-1-yl)acrylic acid (2) were isolated and evaluated for their antimicrobial activities. Their structures were elucidated by UV, IR, MS, 1H-NMR, 13C-NMR and 2D NMR spectra. Compound 1 exhibited the significant activities in vitro against two strains of bacteria and four strains of fungi. Compound 2 gave slight activities on the fungi at 100 µg mL(-1), but no activities on the bacteria. Compound 1 should be considered as a new lead or model compound to develop new isocoumarin antimicrobial agents.
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Pyrrolidine, ≥99.0%