MilliporeSigma
  • Mutagenic/recombinogenic effects of four lipid peroxidation products in Drosophila.
Mutagenic/recombinogenic effects of four lipid peroxidation products in Drosophila.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-12-15)
Eşref Demir, Fatma Turna, Bülent Kaya, Amadeu Creus, Ricard Marcos
ABSTRACT
The human diet is an important factor in the development of different diseases. Lipid peroxidation during frying in edible vegetable liquid oils of food components is a mechanism leading to the formation of free radicals. Such radicals induce tissue damage and are implicated in diverse pathological conditions, including aging, atherosclerosis, brain disorders, cancer, lung disorders and various liver disorders. In the present study, we decided to investigate the genotoxic effects of four lipid peroxidation products in the in vivo Drosophila wing somatic mutation and recombination test. In this test, point mutation, chromosome breakage and mitotic recombination produce single spots; while twin spots are produced only by mitotic recombination. Drosophila is a suitable eukaryotic organism for mutagenicity studies and also its metabolism is quite similar to that of mammalians. Since conflicting data exist on the possible risk of several lipid peroxidation products for humans, we have selected four of them, namely acrolein, crotonaldehyde, 4-hydroxy-hexenal (4-HHE) and 4-oxo-2-nonenal (4-ONE). Especially at the highest concentrations tested all exert both mutagenic and recombinogenic effects in the Drosophila SMART assay, showing a direct dose-effect relationship. This is the first study reporting genotoxicity data in Drosophila for these compounds.
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)