MilliporeSigma
  • Antiaging, antistress and ROS scavenging activity of crude extract of Ocimum sanctum (L.) in Caenorhabditis elegans (Maupas, 1900).
Antiaging, antistress and ROS scavenging activity of crude extract of Ocimum sanctum (L.) in Caenorhabditis elegans (Maupas, 1900).
Indian journal of experimental biology (2013-08-01)
Rakesh Pandey, Shipra Gupta, Virendra Shukla, Sudeep Tandon, Vibha Shukla
ABSTRACT
Since aging is the most important risk factor for variety of diseases, the discovery of a wide range of chemical modulators of aging in model organisms encourages new strategies for targeting age associated diseases. Simple genetic manipulation leads to long-lived and healthy animals, so any compound which could have similar effect would prove a boon to mankind. In the present study, effect of different pharmacological doses (1.0, 0.1, 0.01 and 0.001 mg/mL) of O. sanctum crude extract were used to determine their impact on life span, thermotolerance and ROS scavenging activities in C. elegans. The results revealed that 1 mg/mL of O. sanctum extract significantly extended the life span of C. elegans. The extract also proved to be a strong free radical scavenger and increased resistance against thermal stress. It is also suggested that the protective and life span extending action of the crude extract is not only due to its antioxidant capacity but may also be mediated by modulation of some signaling pathways. Thus, in addition to all the known medicinal property of Ocimum, it is capable of increasing stress tolerance and life span in C. elegans.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)