MilliporeSigma
  • Identification of causative compounds and microorganisms for musty odor occurrence in the Huangpu River, China.
Identification of causative compounds and microorganisms for musty odor occurrence in the Huangpu River, China.
Journal of environmental sciences (China) (2013-08-08)
Daolin Sun, Jianwei Yu, Wei An, Min Yang, Guoguang Chen, Shujun Zhang
ABSTRACT
There are regular problems of musty odor in the Huangpu River, a major source of drinking water for Shanghai, China. In this study, the musty odor and its main causative compounds in the Huangpu River source water were confirmed through a yearly investigation using flavor profile analysis combined with HSPME-GC-MS analysis. The investigation showed that 2-methylisoborneol (2-MIB) with a concentration level between 28.6 and 71.0 ng/L was responsible for the musty odor in summer from July to September. Microscopic observation confirmed with the cloning results showed that Phormidium spp., which accounted for 80%-95% of the algal cell density, was the microorganisms responsible for the production of 2-MIB and the estimated 2-MIB yield was 0.022 pg/cell. Results from a wide-area sampling campaign in the Huangpu River watershed showed that, other than the large tributaries receiving water from Tai Lake, several small creeks close to the intake may have contributed most of the 2-MIB and the Phormidium spp. to the Huangpu River source water. This study provides methodology for the investigation of odor causing compounds and microorganisms in river-type source water, and the result will be useful for water quality control in both source water and drinking water.
Product Description
Supelco
Water, for TOC analysis
Pricing and availability is not currently available.
Supelco
Water, suitable for ion chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Water, BioPerformance Certified
Pricing and availability is not currently available.
Supelco
Water, ACS reagent, for ultratrace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pricing and availability is not currently available.
Sigma-Aldrich
Distilled water, suitable for HPLC
Pricing and availability is not currently available.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pricing and availability is not currently available.
Sigma-Aldrich
Water, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Pricing and availability is not currently available.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Water, HPLC Plus
Pricing and availability is not currently available.
Sigma-Aldrich
Water, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Deionized
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methylisoborneol, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)