MilliporeSigma
  • Inactivation of dihydropteridine reductase (human brain) by platinum(II) complexes.
Inactivation of dihydropteridine reductase (human brain) by platinum(II) complexes.
European journal of biochemistry (1987-04-15)
W L Armarego, A Ohnishi
ABSTRACT
Potassium tetrachloroplatinate (K2PtCl4) inactivates dihydropteridine reductase from human brain in a time-dependent and irreversible manner. The inactivation has been followed by measuring enzyme activity and fluorescence changes. The enzyme is completely protected from inactivation by NADH, the pterin cofactor [quinonoid 6-methyl-7,8-dihydro(6H)pterin] and dithiothreitol. Evidence is presented that K2PtCl4 reacts at the active site and that (a) thiol group(s) is involved in, or is masked by, this reaction. K2PtCl4 is a stronger inhibitor of human brain dihydropteridine reductase that cis- and trans-diaminodichloroplatinum, cis-dichloro[ethylenediamine]platinum and K4Fe(CN)6, whereas H2PtCl6 is considerably weaker and (Ph3P)3RhCl is inactive.
Sigma-Aldrich
Dichloro(ethylenediamine)platinum(II), 99%