Merck
  • The effects of Kv1.3 and IKCa1 potassium channel inhibition on calcium influx of human peripheral T lymphocytes in rheumatoid arthritis.
The effects of Kv1.3 and IKCa1 potassium channel inhibition on calcium influx of human peripheral T lymphocytes in rheumatoid arthritis.
Immunobiology (2012-06-19)
Gergely Toldi, Anna Bajnok, Diána Dobi, Ambrus Kaposi, László Kovács, Barna Vásárhelyi, Attila Balog
ABSTRACT
The transient increase of the cytoplasmic free calcium level plays a key role in the process of lymphocyte activation. Kv1.3 and IKCa1 potassium channels are important regulators of the maintenance of calcium influx during lymphocyte activation and present a possible target for selective immunomodulation. Case-control study. We took peripheral blood samples from 10 healthy individuals and 9 recently diagnosed rheumatoid arthritis (RA) patients receiving no anti-rheumatic treatment. We evaluated calcium influx kinetics following activation in CD4, Th1, Th2 and CD8 cells applying a novel flow cytometry approach. We also assessed the sensitivity of the above subsets to specific inhibition of the Kv1.3 and IKCa1 potassium channels. The peak of calcium influx in lymphocytes isolated from RA patients is reached more rapidly, indicating that they respond more quickly to stimulation compared to controls. In healthy individuals, the inhibition of the IKCa1 channel decreased calcium influx in Th2 and CD4 cells to a lower extent than in Th1 and CD8 cells. On the contrary, the inhibition of Kv1.3 channels resulted in a larger decrease of calcium entry in Th2 and CD4 than in Th1 and CD8 cells. No difference was detected between Th1 and Th2 or CD4 and CD8 cells in the sensitivity to IKCa1 channel inhibition among lymphocytes of RA patients. However, specific inhibition of the Kv1.3 channel acts differentially on calcium influx kinetics in RA lymphocyte subsets. Th2 and particularly CD8 cells are inhibited more dominantly than Th1 and CD4 cells. The inhibition of Kv1.3 channels does not seem to be specific enough in peripheral RA lymphocytes, since anti-inflammatory Th2 cells are also affected to a noteworthy extent.
Sigma-Aldrich
Margatoxin