MilliporeSigma
  • Regeneration of cirrhotic liver in rabbits after intrahepatic injection of cryoprecipitate.
Regeneration of cirrhotic liver in rabbits after intrahepatic injection of cryoprecipitate.
Bulletin of experimental biology and medicine (2013-03-14)
A F Chernousov, T V Khorobrykh, R V Karpova, T P Nekrasova
ABSTRACT
The stimulatory effect of cryoprecipitate on liver regeneration was studied in rabbits with cirrhosis of the liver induced by subcutaneous injection of tetrachloromethane. Cryoprecipitate stimulated the hepatocyte regeneration (appearance of binuclear and proliferating cells) in liver tissue in cirrhosis, but the initial structure of the liver was not restored. The newly formed focus of regeneration with proper girder structure of hepatocytes forced back the connective tissue. Highly concentrated solution of fibrinogen, injected by puncture method into cirrhotic hepatic tissue, stimulated its regeneration.
Product Description
Supelco
Carbon tetrachloride, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
Carbon tetrachloride, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Carbon tetrachloride, reagent grade, 99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46