MilliporeSigma
  • Performance evaluation and modeling of synthetic-fiber barrier in the treatment of turbid water.
Performance evaluation and modeling of synthetic-fiber barrier in the treatment of turbid water.
Water environment research : a research publication of the Water Environment Federation (2013-08-16)
Jianghua Yu, Youngchul Kim
ABSTRACT
A synthetic-fiber barrier for the removal of turbidity in water was developed and tested using a laboratory scale channel. The effects of hydraulics (flow rate and exchange rate); density current caused by temperature and turbidity difference; barrier conditions (thickness, number and shape); and particle size were analyzed. The experimental results indicated that removal efficiency was positively related to barrier thickness and number, was inversely related to the strength of the density current, and was also negatively affected by the flow rate and exchange rate. A wedged barrier was found to work better than a rectangular one when the same amount of fiber was used. Based on the experimental work, empirical models for the removal efficiency and barrier design were established using dimensionless groups. The modeling results indicated that the predicted values were consistent with the experimental work and the increases and decreases in the performance were suitably simulated.
Product Description
Supelco
Water, for TOC analysis
Pricing and availability is not currently available.
Supelco
Water, suitable for ion chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Water, BioPerformance Certified
Pricing and availability is not currently available.
Supelco
Water, ACS reagent, for ultratrace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pricing and availability is not currently available.
Sigma-Aldrich
Distilled water, suitable for HPLC
Pricing and availability is not currently available.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pricing and availability is not currently available.
Sigma-Aldrich
Water, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Pricing and availability is not currently available.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Water, HPLC Plus
Pricing and availability is not currently available.
Sigma-Aldrich
Water, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Deionized
Pricing and availability is not currently available.