MilliporeSigma
Analyses of two azo dyes by high-performance liquid chromatography.
Journal of chromatographic science (1988-09-01)
A Shan, D Harbin, C W Jameson
ABSTRACT
Two structurally similar azo dyes, Direct Blue No. 6 and Direct Blue No. 15, are analyzed by separate reversed-phase high-performance liquid chromatographic (HPLC) systems utilizing a dual-wavelength UV/visible detector at 254 and 546 nm. Each dye contains more than 35 impurity peaks. Different lots of each dye are compared by means of their respective chromatographic profiles. The system used to obtain a chromatographic profile for each dye is adapted to make comparisons of the major component in different lots by an internal standard method. These methods are reproducible, giving relative standard deviations ranging from approximately 1 to 5%. These HPLC systems are adaptable for the analysis of other structurally similar dyes.
Sigma-Aldrich
Direct Blue 15, suitable for Histopaque® system, suitable for viability studies of collagenase-treated rat liver cells