MilliporeSigma
  • Accumulation of 1-methyl-tetrahydro-beta-carboline-3-carboxylic acid in blood and organs of rat. A possible causative substance of eosinophilia-myalgia syndrome associated with ingestion of L-tryptophan.
Accumulation of 1-methyl-tetrahydro-beta-carboline-3-carboxylic acid in blood and organs of rat. A possible causative substance of eosinophilia-myalgia syndrome associated with ingestion of L-tryptophan.
Archives of toxicology (1993-01-01)
Y Ogawa, J Adachi, Y Tatsuno
ABSTRACT
1-Methyl-tetrahydro-beta-carboline-3-carboxylic acid (MTCA) may cause eosinophilia-myalgia syndrome (EMS) associated with ingestion of L-tryptophan. The distribution and excretion of MTCA were studied in rats which had received perorally a single 1.6 mg/kg dose of MTCA. MTCA concentrations in blood, kidney, liver, brain, heart, spleen, lung and gastrocnemius muscle were measured by HPLC combined with fluorometric detection. The concentration of MTCA in each organ reached a maximum at 1 h and then gradually declined. However, a significant level of MTCA still remained at 5 h, when 52% of ingested MTCA remained in the contents of the large intestine. Twenty-nine percent of the ingested MTCA was excreted in urine over the course of 24 h. A higher dose (10 mg/kg) of MTCA resulted in significant elevations in the concentrations and amounts of MTCA in the various organs. In addition, chronic treatment with a 10 mg/kg dose of MTCA for 6 weeks further increased the concentrations and amounts of MTCA in each organ. However, no histological changes were observed in any of the organs after chronic treatment. This is the first report which demonstrates accumulation of MTCA in the blood and various organs, including muscle, of rats.
Sigma-Aldrich
1,2,3,4-Tetrahydroharmane-3-carboxylic acid