MilliporeSigma
  • Measurement of immune markers in the serum and cerebrospinal fluid of multiple sclerosis patients during clinical remission.
Measurement of immune markers in the serum and cerebrospinal fluid of multiple sclerosis patients during clinical remission.
Journal of neurology (1995-01-01)
C E Shaw, P R Dunbar, H A Macaulay, T J Neale
ABSTRACT
Magnetic resonance imaging of multiple sclerosis (MS) patients often shows active inflammatory lesions despite clinical remission. No immunological marker of disease activity has been identified in these patients. Concentrations of neopterin, interleukin-2 (IL-2), soluble interleukin-2 receptor (sIL-2R) and tumour necrosis factor-alpha (TNF-alpha) were measured in the serum and cerebrospinal fluid of 19 clinically-inactive MS patients and compared with those of 19 non-inflammatory controls. Cerebrospinal fluid (CSF) neopterin concentrations were significantly higher in the MS group than in controls (mean 9.1 mM vs 3.4 nM, P < 0.01) and 10 of 19 MS patients had levels above the control range. This finding provides evidence of ongoing T-cell-directed and interferon-gamma-mediated macrophage activation in the central nervous system. Analysis of IL-2, sIL-2R and TNF-alpha concentrations revealed no significant differences between MS patients and controls. We conclude that CSF neopterin concentration may correlate with disease activity in asymptomatic patients.
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Mercury, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Mercury, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis