MilliporeSigma
  • Correction of water content and solute concentration in blood during hemoconcentration.
Correction of water content and solute concentration in blood during hemoconcentration.
Journal of applied physiology: respiratory, environmental and exercise physiology (1977-05-01)
Y Ohira, A Ito, S Ikawa
PMID863843
ABSTRACT
Eleven, healthy male students were subjected to an exhaustive bicyle ergometer exercise. Many parameters of hemoconcentration were examined in blood and urine samples which were collected before and after exercise. In an exercise lasting approximately 10 min, it was found that serum total protein (STP) would be a suitable parameter that could be used to calculate water content and actual decrease in water volume in blood due to exercise. Further, it is described how STP values before and during hemoconcentration can be used to eliminate the effect of plasma water loss from the determined values of various solute levels in blood.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Distilled water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
Water, HPLC Plus
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00