MilliporeSigma
  • Response to a long-term high-fat diet in the signature of genes involved in lipid metabolism in ApoE(-/-) mice.
Response to a long-term high-fat diet in the signature of genes involved in lipid metabolism in ApoE(-/-) mice.
The American journal of the medical sciences (2012-11-10)
Mengliu Yang, Shengbing Li, Hua Liu, Gangyi Yang, Ling Li
ABSTRACT
High-fat diet (HFD)-induced lipid metabolism disorders are a critical feature of diet-induced insulin resistance (IR); however, the mechanisms underlying the tissue-specific effects of HFDs on the signature of genes involved in lipid metabolism have not been fully investigated. Glucose homeostasis and insulin sensitivity were monitored in chow-fed and HFD-fed ApoE mice using the hyperinsulinemic-euglycemic clamp technique. The signatures of genes involved in lipid metabolism were measured in the liver and adipose tissues by quantitative real-time polymerase chain reaction and Western blot analysis. Long-term HFD (for 16 weeks) resulted in marked abnormalities in glucose and lipid metabolism and induced IR in ApoE mice. Long-term HFD also markedly upregulated the messenger RNA (mRNA) expression of insulin-induced gene-2, sterol regulatory element-binding protein cleavage-activating protein and sterol regulatory element-binding protein-1 in the liver and/or adipose tissues, in parallel with an elevated insulin-induced gene-2 protein in the liver. However, HFD markedly downregulated the mRNA expression of sterol regulatory element-binding protein-2, 3-hydroxy-3-methylglutaryl coenzyme A reductase and low-density lipoprotein receptor in the liver. Interestingly, long-term HFD markedly decreased adipose triglyceride lipase and peroxisome proliferator-activated receptor-γ mRNA, and adipose triglyceride lipase protein contents in the liver and/or adipose tissues. These findings provide a framework to understand the mechanisms by which long-term HFD regulates the gene signatures involved in lipid metabolism in the pathogenesis of diet-induced IR.
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g